COMH4211BF7E Combi-Blocks, Inc.

Benzenemethanamine, 3-bromo-4-(phenylmethoxy)-, hydrochloride

  • MDL number MFCD28383615
  • Cas Number : 375854-71-0
  • Molecular Weight : 328.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7133
Availability details for
Quantity
  • Availability
  • Add to Cart