COMH4211BF7F Combi-Blocks, Inc.

3-(2-Fluoro-4-methyl-phenyl)-2-methyl-propionic acid

  • MDL number MFCD26639658
  • Cas Number : 1513229-84-9
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7134
Availability details for
Quantity
  • Availability
  • Add to Cart