COMH4211BF80 Combi-Blocks, Inc.

C-(4-Chloro-3-fluoro-phenyl)-c-phenyl-methylamine hydrochloride

  • MDL number MFCD31557824
  • Cas Number : 2206265-88-3
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7135
Availability details for
Quantity
  • Availability
  • Add to Cart