COMH4211BF81 Combi-Blocks, Inc.

Ethyl 6-(cyclopropylsulfonyl)nicotinate

  • MDL number MFCD27937144
  • Cas Number : 1993309-64-0
  • Molecular Weight : 255.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 6-(cyclopropylsulfonyl)nicotinate
Inchi Code 1S/C11H13NO4S/c1-2-16-11(13)8-3-6-10(12-7-8)17(14,15)9-4-5-9/h3,6-7,9H,2,4-5H2,1H3
Inchi Key IVZSXHQUCRXFNO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7136
Availability details for
Quantity
  • Availability
  • Add to Cart