COMH4211BFDE Combi-Blocks, Inc.

1-Cyclopropanesulfonyl-1h-pyrazol-4-ol

  • MDL number MFCD31579526
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7253
Availability details for
Quantity
  • Availability
  • Add to Cart