COMH4211BFE3 Combi-Blocks, Inc.

(2S)-1-(Propane-2-sulfonyl)piperidine-2-carboxylic acid

  • MDL number MFCD21733068
  • Cas Number : 1567929-74-1
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7261
Availability details for
Quantity
  • Availability
  • Add to Cart