COMH4211BFE8 Combi-Blocks, Inc.

1-(3-Chloro-4-(trifluoromethoxy)benzyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD31583089
  • Cas Number : 1604036-66-9
  • Molecular Weight : 402.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7275

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7275
Availability details for
Quantity
  • Availability
  • Add to Cart