COMH4211BFE9 Combi-Blocks, Inc.

6-(4-Bromobutoxy)benzo[d]thiazole

  • MDL number MFCD31613706
  • Cas Number : 1953183-92-0
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7282
Availability details for
Quantity
  • Availability
  • Add to Cart