COMH4211BFEC Combi-Blocks, Inc.

1-(2-Isopropoxyethyl)-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD22571844
  • Cas Number : 1339890-98-0
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7286
Availability details for
Quantity
  • Availability
  • Add to Cart