COMH4211BFFA Combi-Blocks, Inc.

Benzyl 2-iodophenyl(methyl)carbamate

  • MDL number MFCD31613851
  • Cas Number : 1860876-40-9
  • Molecular Weight : 367.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7312
Availability details for
Quantity
  • Availability
  • Add to Cart