COMH4211C004 Combi-Blocks, Inc.

2-Bromo-5-(pyrrolidine-1-carbonyl)pyridine

  • MDL number MFCD14702611
  • Cas Number : 1209460-17-2
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7334

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7334
Availability details for
Quantity
  • Availability
  • Add to Cart