COMH4211C005 Combi-Blocks, Inc.

1-Bromo-2-fluoro-4-(2-methylpropoxy)benzene

  • MDL number MFCD21142825
  • Cas Number : 1496272-53-7
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7335
Availability details for
Quantity
  • Availability
  • Add to Cart