COMH4211C016 Combi-Blocks, Inc.

[1,1'-Biphenyl]-4-carboxylic acid, 4'-fluoro-2-hydroxy-, methyl ester

  • MDL number MFCD31381614
  • Cas Number : 198994-01-3
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7381

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7381
Availability details for
Quantity
  • Availability
  • Add to Cart