COMH4211C027 Combi-Blocks, Inc.

4-((Cyclopropylamino)methyl)-2-fluorophenol

  • MDL number MFCD26674997
  • Cas Number : 1537583-94-0
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7420
Availability details for
Quantity
  • Availability
  • Add to Cart