COMH4211C042 Combi-Blocks, Inc.

Desthiobiotin n-hydroxysuccinimidyl ester

  • MDL number MFCD08062086
  • Cas Number : 80750-24-9
  • Molecular Weight : 311.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7432
Availability details for
Quantity
  • Availability
  • Add to Cart