COMH4211C049 Combi-Blocks, Inc.

3-(Propan-2-ylsulfanyl)propanoic acid

  • MDL number MFCD09938047
  • Cas Number : 24383-50-4
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7459
Availability details for
Quantity
  • Availability
  • Add to Cart