COMH4211C05F Combi-Blocks, Inc.

3-N-Butoxy-5-fluorobenzoic acid

  • MDL number MFCD18426789
  • Cas Number : 1443311-12-3
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7517
Availability details for
Quantity
  • Availability
  • Add to Cart