COMH4211C077 Combi-Blocks, Inc.

1-(2-Allyloxy-ethyl)-4-bromo-benzene

  • MDL number MFCD31619000
  • Cas Number : 197633-71-9
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7573

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7573
Availability details for
Quantity
  • Availability
  • Add to Cart