COMH4211C078 Combi-Blocks, Inc.

2-Amino-5-pyrazol-1-yl-benzoic acid methyl ester

  • MDL number MFCD14621767
  • Cas Number : 1219950-35-2
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7574
Availability details for
Quantity
  • Availability
  • Add to Cart