COMH4211C087 Combi-Blocks, Inc.

2-Chloro-5-cyclopropylbenzoic acid

  • MDL number MFCD01070422
  • Cas Number : 1119088-88-8
  • Molecular Weight : 196.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7598
Availability details for
Quantity
  • Availability
  • Add to Cart