COMH4211C0B1 Combi-Blocks, Inc.

3-(4-Methylpiperazin-1-yl)phenol

  • MDL number MFCD12405778
  • Cas Number : 177489-10-0
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7651

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7651
Availability details for
Quantity
  • Availability
  • Add to Cart