COMH4211C0C8 Combi-Blocks, Inc.

2-[1-(Cyclopropanesulfonyl)-1h-pyrazol-4-yl]pyrimidin-4-amine

  • MDL number MFCD30803782
  • Cas Number : 1612174-02-3
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7700
Availability details for
Quantity
  • Availability
  • Add to Cart