COMH4211C0DC Combi-Blocks, Inc.

3,5-Difluoro-4-isobutoxybenzoic acid

  • MDL number MFCD18285386
  • Cas Number : 1343670-92-7
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7743
Availability details for
Quantity
  • Availability
  • Add to Cart