COMH4211C0F5 Combi-Blocks, Inc.

3-(4-Hydroxy-phenoxymethyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583290
  • Molecular Weight : 307.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7766
Availability details for
Quantity
  • Availability
  • Add to Cart