COMH4211C0FA Combi-Blocks, Inc.

D-Valine, n-[(1,1-dimethylethoxy)carbonyl]-l-alanyl-

  • MDL number MFCD31381710
  • Cas Number : 139601-17-5
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7775
Availability details for
Quantity
  • Availability
  • Add to Cart