COMH4211C0FB Combi-Blocks, Inc.

3-Fluoro-4-hydroxy-benzoic acid tert-butyl ester

  • MDL number MFCD31613824
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7781
Availability details for
Quantity
  • Availability
  • Add to Cart