COMH4211C10D Combi-Blocks, Inc.

5-Phenoxypentanal

  • MDL number MFCD17252877
  • Cas Number : 87841-94-9
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7823

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7823
Availability details for
Quantity
  • Availability
  • Add to Cart