COMH4211C120 Combi-Blocks, Inc.

2-(Isopropylthio)-5-nitrobenzaldehyde

  • MDL number MFCD18887928
  • Cas Number : 900174-36-9
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7854
Availability details for
Quantity
  • Availability
  • Add to Cart