COMH4211C124 Combi-Blocks, Inc.

1H-Pyrazole-1-acetic acid, alpha,alpha-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1,1-dimethylethyl ester

  • MDL number MFCD22571828
  • Cas Number : 1006876-29-4
  • Molecular Weight : 336.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7861

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7861
Availability details for
Quantity
  • Availability
  • Add to Cart