COMH4211C126 Combi-Blocks, Inc.

1-(2, 3-Difluorobenzyl)-1h-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD31579370
  • Cas Number : 1996875-29-6
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7864
Availability details for
Quantity
  • Availability
  • Add to Cart