COMH4211C132 Combi-Blocks, Inc.

2-tert-Butyl-4-chloroaniline

  • MDL number MFCD20726042
  • Cas Number : 959528-03-1
  • Molecular Weight : 183.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7907

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7907
Availability details for
Quantity
  • Availability
  • Add to Cart