COMH4211C13E Combi-Blocks, Inc.

5-Bromo-1h-indole-3-carboxamide

  • MDL number MFCD31583246
  • Cas Number : 1928780-39-5
  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7936

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7936
Availability details for
Quantity
  • Availability
  • Add to Cart