COMH4211C157 Combi-Blocks, Inc.

tert-Butyl-(2-(3-iodophenoxy)ethoxy)dimethylsilane

  • MDL number MFCD31583140
  • Cas Number : 940876-04-0
  • Molecular Weight : 378.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7954
Availability details for
Quantity
  • Availability
  • Add to Cart