COMH4211C161 Combi-Blocks, Inc.

2-Methoxy-n-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene sulfonamide

  • MDL number MFCD29109273
  • Cas Number : 1534377-89-3
  • Molecular Weight : 327.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7974

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7974
Availability details for
Quantity
  • Availability
  • Add to Cart