COMH4211C162 Combi-Blocks, Inc.

3-(4-Hydroxymethylphenyl)butyric acid ethyl ester

  • MDL number MFCD17018302
  • Cas Number : 862855-56-9
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7975
Availability details for
Quantity
  • Availability
  • Add to Cart