COMH4211C170 Combi-Blocks, Inc.

1-(2,2,2-Trifluoroethyl)pyrrolidin-3-ol

  • MDL number MFCD17248893
  • Molecular Weight : 169.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8003
Availability details for
Quantity
  • Availability
  • Add to Cart