COMH4211C172 Combi-Blocks, Inc.

3-(2-Methoxy-ethanesulfonyl)-propionic acid

  • MDL number MFCD09947463
  • Cas Number : 1016858-81-3
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8011
Availability details for
Quantity
  • Availability
  • Add to Cart