COMH4211C17B Combi-Blocks, Inc.

1-Benzyl-4-(2h-tetrazol-5-yl)-1h-indole

  • MDL number MFCD31579419
  • Cas Number : 177548-79-7
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8027
Availability details for
Quantity
  • Availability
  • Add to Cart