COMH4211C189 Combi-Blocks, Inc.

4-(6-Oxo-1,6-dihydro-pyrimidin-4-yl)-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31613714
  • Cas Number : 1159568-09-8
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8070

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8070
Availability details for
Quantity
  • Availability
  • Add to Cart