COMH4211C192 Combi-Blocks, Inc.

1-(4-Methoxy-2-methyl-phenyl)-propylamine hydrochloride

  • MDL number MFCD31557834
  • Cas Number : 2205505-33-3
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8086
Availability details for
Quantity
  • Availability
  • Add to Cart