COMH4211C1BA Combi-Blocks, Inc.

3-(4-Chloro-2-fluoro-phenyl)-5-methyl-isoxazole-4-carboxylic acid ethyl ester

  • MDL number MFCD22370423
  • Cas Number : 1159602-71-7
  • Molecular Weight : 283.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8123
Availability details for
Quantity
  • Availability
  • Add to Cart