COMH4211C1F4 Combi-Blocks, Inc.

(Z)-N-Methoxy-n-methyloctadec-9-enamide

  • MDL number MFCD31613922
  • Cas Number : 190131-27-2
  • Molecular Weight : 325.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8247
Availability details for
Quantity
  • Availability
  • Add to Cart