COMH4211C21E Combi-Blocks, Inc.

1-(Cyclohexylmethyl)piperidin-4-ol

  • MDL number MFCD13514838
  • Cas Number : 149398-34-5
  • Molecular Weight : 197.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8289
Availability details for
Quantity
  • Availability
  • Add to Cart