COMH4211C220 Combi-Blocks, Inc.

3-Bromo-4-(3-fluoropropoxy)-5-methoxybenzaldehyde

  • MDL number MFCD24049232
  • Cas Number : 1514790-88-5
  • Molecular Weight : 291.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8294

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8294
Availability details for
Quantity
  • Availability
  • Add to Cart