COMH4211C225 Combi-Blocks, Inc.

3-[2-(2-Propen-1-yloxy)ethoxy]-propionic acid

  • MDL number MFCD20643582
  • Cas Number : 13630-56-3
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8308
Availability details for
Quantity
  • Availability
  • Add to Cart