COMH4211C24C Combi-Blocks, Inc.

(2-Bromo-4-methyl-benzyl)-cyclopropyl-amine hydrochloride

  • MDL number MFCD31613862
  • Molecular Weight : 276.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8384
Availability details for
Quantity
  • Availability
  • Add to Cart