COMH4211C252 Combi-Blocks, Inc.

Methyl 2-fluoro-6-(pyrimidin-2-yl)benzoate

  • MDL number MFCD26401352
  • Cas Number : 1293284-62-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8401

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8401
Availability details for
Quantity
  • Availability
  • Add to Cart