COMH4211C254 Combi-Blocks, Inc.

4-Amino-2-(cyclopropylmethoxy)-n-methylbenzamide

  • MDL number MFCD19640853
  • Cas Number : 1342482-23-8
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8408

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8408
Availability details for
Quantity
  • Availability
  • Add to Cart