COMH4211C27C Combi-Blocks, Inc.

(2-Amino-5-fluorophenyl)(morpholino)methanone

  • MDL number MFCD11132584
  • Cas Number : 1094449-18-9
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8445
Availability details for
Quantity
  • Availability
  • Add to Cart