COMH4211C28D Combi-Blocks, Inc.

Pyrrolidin-1-yl-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-yl]-methanone

  • MDL number MFCD31583372
  • Molecular Weight : 291.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8477
Availability details for
Quantity
  • Availability
  • Add to Cart